Learn More
Strem, An Ascensus Company CAS# 64-02-8. 500g. Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99+%. MFCD00150027
SDP
Catalog No. 5090113833
Encompass
CAS# 64-02-8. 500g. Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99+%. MFCD00150027. Molecular Weight: 380.18 (452.23). Molecular Formula: (NaOOCCH2)2NCH2CH2N(CH2COONa)2·4H2O. Color/form: white xtl. Strem# 93-1104. http://www.strem.com/catalog/v/93-1104/
Catalog No. 50-901-13833 Supplier Strem, An Ascensus Company Supplier No. 931104500G
CAS# 64-02-8. 500g. Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99+%. MFCD00150027. Molecular Weight: 380.18 (452.23). Molecular Formula: (NaOOCCH2)2NCH2CH2N(CH2COONa)2·4H2O. Color/form: white xtl. Strem# 93-1104. http://www.strem.com/catalog/v/93-1104/