Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediaminetetraacetic acid solution, 60-00-4, MFCD00003541, 100mL
Catalog No. 501657060
Encompass
Empirical Formula (Hill Notation) C10H16N2O8, Molecular Weight 292.24, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O, Synonym: EDTA solution
Catalog No. 50-165-7060 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 03690100ML
Empirical Formula (Hill Notation) C10H16N2O8, Molecular Weight 292.24, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O, Synonym: EDTA solution