Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid | 60-00-4 | >99.0% | 292.24 g/mol | C10H16N2O8 | 25g
SDP
Catalog No. 5000267855
Encompass_Preferred
Ethylenediaminetetraacetic acid (EDTA) is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid can alleviate the liver fibrosis Ethylenediaminetetraacetic acid can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]
Catalog No. 50-002-67855 Supplier Medchemexpress LLC Supplier No. HYY068225G
Ethylenediaminetetraacetic acid (EDTA) is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid can alleviate the liver fibrosis Ethylenediaminetetraacetic acid can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]