Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid | 60-00-4 | MFCD00003541 | ≥98.0% | 292.24 g/mol | C10H16N2O8 | 100g
SDP
Supplier:  Medchemexpress LLC HYY0682100G
Encompass_Preferred
Ethylenediaminetetraacetic acid (EDTA) is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid can alleviate the liver fibrosis Ethylenediaminetetraacetic acid can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]
Catalog No. 50-002-68619