Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylenediaminetetraacetic Acid Calcium Disodium Salt Hydrate

Catalog No. 340073100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-007-3100G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 34-007-3100G Supplier Sigma Aldrich Supplier No. 340073100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylenediaminetetraacetic acid calcium disodium salt hydrate is calcium and sodium salt of EDTA and it can be used:To retain the quality of litchi fruit by the postharvest treatment process.In the preparation of an aqueous solution for hydroxyapatite coating, which is used to coat the magnesium material/particle for biomedical applications.To prepare a mobile phase for the separation of diol products from the aqueous solution of the reaction mixture by chromatography techniques.

Specifications

CAS 223719-29-7
Boiling Point 190°C to 195°C (0.05 mmHg, lit.)
Molecular Formula C28H52O6
Linear Formula C28H52O6
MDL Number MFCD00068562
Quantity 5 g
Molecular Weight (g/mol) 484.71 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.