Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid, Di K Salt Dihydrate (White Crystalline Powd.), Fisher BioReagents

Catalog No. BP119500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP119-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP119-500

missing translation for 'validMainframeMsg'

The dipotassium salt of EDTA is used in place of the disodium salt where sodium would interfere with the reaction.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25102-12-9
Molecular Formula C10H18K2N2O10
Molecular Weight (g/mol) 404.455
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx
PubChem CID 53486451
IUPAC Name dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+]

Specifications

Melting Point 272°C
Color White
pH 4.0 to 5.0
Assay Percent Range ≥99.0 %
Packaging Glass Bottle
Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOK)2·2H2O
Quantity 500 g
Formula Weight 404.47
Percent Purity ≥99.0%
Physical Form Powder or Solid
Chemical Name or Material Ethylenediamine Tetraacetic Acid, Dipotassium Salt Dihydrate
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.