Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic acid diammonium copper salt Solution, volumetric, 0.025 M EDTA-Cu(NH4)2, MilliporeSigma™ Supelco™

Catalog No. 111015150 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5150 1 L
11-101-5151 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-5150 Supplier MilliporeSigma Supelco Supplier No. 461221LF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Visit our Sensor Applications portal to learn more.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD01863036
Synonym Copper diammonium EDTA; Diammonium copper ethylenediaminetetraacetate solution

Specifications

CAS 105512-06-9
Linear Formula C10H20 CuN4O8
Quantity 1 L
Formula Weight 387.83
Concentration or Composition (by Analyte or Components) 0.025 M EDTA-Cu(NH4)2
ShelfLife Limited shelf life, expiry date on the label
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.