Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid dihydrate tripotassium | 65501-24-8 | MFCD00149672 | 442.54 | C10H17K3N2O10 | 25 G
SDP
Catalog No. 5000209590
Encompass_Preferred
EDTA·2H2O (tripotassium) is the tripotassium dihydrate salt of ethylenediaminetetraacetic acid, supplied as a solid chelating reagent for life-science research. It functions as a metal-ion chelator and is provided in laboratory-scale packages with recommended storage for both powder and solution forms.
  • Metal-ion chelator effective against divalent and trivalent cations.
  • Provided as a solid, typically a white crystalline powder.
  • Available in laboratory pack sizes suitable for routine use, such as 25 G.
  • Storage recommendations: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); solutions stable at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Common applications include buffer preparation, metal-ion sequestration, and use as an anticoagulant/chelating agent in biochemical assays.
Catalog No. 50-002-09590 Supplier Medchemexpress LLC Supplier No. 25GHYW087947
EDTA·2H2O (tripotassium) is the tripotassium dihydrate salt of ethylenediaminetetraacetic acid, supplied as a solid chelating reagent for life-science research. It functions as a metal-ion chelator and is provided in laboratory-scale packages with recommended storage for both powder and solution forms.
  • Metal-ion chelator effective against divalent and trivalent cations.
  • Provided as a solid, typically a white crystalline powder.
  • Available in laboratory pack sizes suitable for routine use, such as 25 G.
  • Storage recommendations: powder stable at -20°C (up to 3 years) or 4°C (up to 2 years); solutions stable at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Common applications include buffer preparation, metal-ion sequestration, and use as an anticoagulant/chelating agent in biochemical assays.