Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%, extra pure

Catalog No. AC209740025
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209740025 2.5 kg Plastic bottle
AC209741000 100 g Plastic bottle
AC209745000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC209740025 Supplier Thermo Scientific Chemicals Supplier No. 209740025
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 25102-12-9 | C10H14K2N2O8·2H2O | 404.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25102-12-9
Molecular Formula C10H14K2N2O8·2H2O
Molecular Weight (g/mol) 404.46
MDL Number MFCD00150036
InChI Key JFROQFOCFOKDKU-UHFFFAOYSA-L
Synonym ethylenediaminetetraacetic acid dipotassium salt dihydrate, dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, potassium 2,2'-2-bis carboxymethyl amino ethyl azanediyl diacetate dihydrate, edta dipotassium salt dihydrate, edta dipotassium salt, ethylenediaminetetraacetic acid,dipotassium salt dihydrate, potassium ethylenediaminetetraacetate dibasic, dipotassium ethylenediaminetetraacetate dihydrate, ethylenedinitrilo tetraacetic acid dipotassium salt, acmc-209ggx
PubChem CID 53486451
IUPAC Name dipotassium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
SMILES C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[K+].[K+]

Specifications

Melting Point 272.0°C
Color White
pH 4 to 5 (5% soln.)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Complexometry)
Packaging Plastic bottle
Linear Formula KO2CCH2N(CH2CO2H)CH2CH2N(CH2CO2K)CH2CO2H
Quantity 2.5 kg
Merck Index 14, 3517
Solubility Information (10% in water) Clear colorless
Formula Weight 404.46
Percent Purity 99%
Grade Extra Pure
Physical Form Fine Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, dipotassium salt dihydrate, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unw

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.