Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylenediaminetetraacetic acid disodium | 139-33-3 | MFCD00070672 | 98.0% | 336.21 g/mol | C10H14N2Na2O8 | 250 MG
SDP

Catalog No. 5000302864
Encompass_Preferred

Ethylenediaminetetraacetic acid disodium is a water-soluble chelating agent and biochemical reagent used to bind metal ions for analytical, laboratory, and industrial applications.

  • High purity, 98.0%, suitable for research and analytical applications.
  • Water-soluble chelating agent for sequestering divalent and trivalent metal ions.
  • Commonly used as a reagent, preservative, stabilizer, and in water treatment processes.
  • Molecular weight 336.21 g/mol; formula C10H14N2Na2O8.
  • Supplied with a safety data sheet and documentation for laboratory use.

Catalog No. 50-003-02864 Supplier Medchemexpress LLC Supplier No. 250MGHYW018746R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylenediaminetetraacetic acid disodium is a water-soluble chelating agent and biochemical reagent used to bind metal ions for analytical, laboratory, and industrial applications.

  • High purity, 98.0%, suitable for research and analytical applications.
  • Water-soluble chelating agent for sequestering divalent and trivalent metal ions.
  • Commonly used as a reagent, preservative, stabilizer, and in water treatment processes.
  • Molecular weight 336.21 g/mol; formula C10H14N2Na2O8.
  • Supplied with a safety data sheet and documentation for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.