Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ambeed AmBeed Ethylenediaminetetraacetic Acid Disodium Dihydrate, 98% Purity, Chemical Reagent, 1 g
SDP

Catalog No. NC3930792
Encompass_Preferred

Ethylenediaminetetraacetic acid disodium dihydrate is a chemical reagent commonly used in laboratory applications.

  • Purity: 98%
  • CAS number: 6381-92-6
  • Storage condition: Sealed in dry, room temperature

Catalog No. NC3930792 Supplier Ambeed Supplier No. A626916100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylenediaminetetraacetic acid disodium dihydrate is a chemical reagent commonly used in laboratory applications.

  • Purity: 98%
  • CAS number: 6381-92-6
  • Storage condition: Sealed in dry, room temperature

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.