Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid disodium dihydrate | 6381-92-6 | MFCD00150037 | 99.9% | 372.24 g/mol | C10H18N2Na2O10 | 25g
SDP
Catalog No. 5000268714
Encompass_Preferred
Ethylenediaminetetraacetic acid (EDTA) disodium dehydrate is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid disodium dehydrate has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid disodium dehydrate decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid disodium dehydrate can alleviate the liver fibrosis Ethylenediaminetetraacetic acid disodium dehydrate can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]
Catalog No. 50-002-68714 Supplier Medchemexpress LLC Supplier No. HYY0682A25G
Ethylenediaminetetraacetic acid (EDTA) disodium dehydrate is a kind of metal chelating agent (binds to bivalent and trivalent metal cations including calcium) Ethylenediaminetetraacetic acid disodium dehydrate has antibacterial anti-inflammatory antioxidant anti-hypercalcemia and anticoagulant activities Ethylenediaminetetraacetic acid disodium dehydrate decreases the metal ion-catalyzed oxidative damage to proteins and allows maintenance of reducing environment during protein purification Ethylenediaminetetraacetic acid disodium dehydrate can alleviate the liver fibrosis Ethylenediaminetetraacetic acid disodium dehydrate can be used for coronary artery disease and neural system disease research[1][2][3][4][5][6][7]