Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylenediaminetetraacetic Acid Disodium Magnesium Salt Hydrate

Catalog No. 317810250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
31-781-0250G 250 g
31-781-050G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 31-781-0250G Supplier Sigma Aldrich Supplier No. 317810250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylenediaminetetraacetic Acid Disodium Magnesium Salt Hydrate

Specifications

CAS 14402-88-1
Molecular Formula C10H12MgN2Na2O8 -+ xH2O
Linear Formula (NaOOCCH2)2NCH2CH2N(CH2COO)2Mg -+ xH2O
MDL Number MFCD03613601
Quantity 250 g
Synonym Ethylenediaminetetraacetic acid disodium magnesium salt; Magnesium disodium ethylenediaminetetraacetate hydrate
Molecular Weight (g/mol) 358.5 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.