Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid disodium salt | 139-33-3 | MFCD00150037 | 336.21 g·mol⁻¹ | C10H14N2Na2O8 | 100 MG
SDP
Catalog No. 5000300707
Encompass_Preferred
EDTA (disodium) is the disodium salt of ethylenediaminetetraacetic acid, a widely used chelating agent that forms stable complexes with divalent and trivalent metal ions. Supplied as a solid reagent for research and analytical applications, it is used to sequester metal contaminants, stabilize samples, and prepare buffered solutions for biochemical and analytical workflows.
  • Chelates calcium and other metal ions to prevent interference in assays.
  • Stabilizes enzyme and protein solutions by removing trace metals.
  • Improves accuracy of analytical measurements by binding metal contaminants.
  • Solid reagent form suitable for precise formulation and storage.
  • Compatible with aqueous buffers and common laboratory procedures.
Catalog No. 50-003-00707 Supplier Medchemexpress LLC Supplier No. HYW018746R100MG
EDTA (disodium) is the disodium salt of ethylenediaminetetraacetic acid, a widely used chelating agent that forms stable complexes with divalent and trivalent metal ions. Supplied as a solid reagent for research and analytical applications, it is used to sequester metal contaminants, stabilize samples, and prepare buffered solutions for biochemical and analytical workflows.
  • Chelates calcium and other metal ions to prevent interference in assays.
  • Stabilizes enzyme and protein solutions by removing trace metals.
  • Improves accuracy of analytical measurements by binding metal contaminants.
  • Solid reagent form suitable for precise formulation and storage.
  • Compatible with aqueous buffers and common laboratory procedures.