Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.2M, Solstice

Catalog No. 6002594 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
Cubitainer
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-025-94 1 L Poly Bottle
60-025-95 5 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-025-94 Supplier Solstice Supplier No. 351021L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.2 M EDTA-Na2, for complexometry

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt solution
Molecular Formula C10H14N2Na2O8
Linear Formula [-CH2N(CH2CO2Na)CH2CO2H]2
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
UN Number NONH for all modes of transport
Beilstein 3822669
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Formula Weight 336.21g/mol
Packaging Poly Bottle
Quantity 1 L
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.