Learn More
Alkali Scientific Ethylenediaminetetraacetic acid [EDTA], 5 Kg
SDP
Catalog No. 5000661876
Encompass_Preferred
EDTA (Ethylenediaminetetraacetic acid) is a powerful chelating agent used extensively in laboratory applications to bind and neutralize metal ions. This 5kg package is ideal for large laboratories or research facilities that require a substantial quantity of EDTA for ongoing experiments. EDTA effectively chelates a variety of metal ions such as calcium, magnesium, and iron, making it indispensable in fields like biochemistry, molecular biology, and environmental science. This reagent is commonly used to prevent metal ion interference in biochemical reactions, enzyme assays, and in the preparation of solutions and buffers where precise metal ion control is necessary for accurate experimental outcomes.
Catalog No. 50-006-61876 Supplier Alkali Scientific Supplier No. DTA011CP
EDTA (Ethylenediaminetetraacetic acid) is a powerful chelating agent used extensively in laboratory applications to bind and neutralize metal ions. This 5kg package is ideal for large laboratories or research facilities that require a substantial quantity of EDTA for ongoing experiments. EDTA effectively chelates a variety of metal ions such as calcium, magnesium, and iron, making it indispensable in fields like biochemistry, molecular biology, and environmental science. This reagent is commonly used to prevent metal ion interference in biochemical reactions, enzyme assays, and in the preparation of solutions and buffers where precise metal ion control is necessary for accurate experimental outcomes.