Learn More
Alkali Scientific Ethylenediaminetetraacetic acid [EDTA], 500 G
SDP
Catalog No. 5000659843
Encompass_Preferred
EDTA (Ethylenediaminetetraacetic acid) is a highly effective chelating agent used for binding metal ions in laboratory applications. This 500g package is designed for smaller-scale experiments where controlling metal ion concentration is critical. EDTA is essential in a variety of fields, including molecular biology, biochemistry, and environmental chemistry, as it prevents metal ions from interfering with biological reactions and analytical procedures. The ability of EDTA to sequester divalent metal ions, particularly calcium and magnesium, makes it indispensable in enzyme assays, protein purification, and DNA/RNA manipulation. It is commonly used in buffer solutions and as a preservative in biological samples.
Catalog No. 50-006-59843 Supplier Alkali Scientific Supplier No. DTA010CP
EDTA (Ethylenediaminetetraacetic acid) is a highly effective chelating agent used for binding metal ions in laboratory applications. This 500g package is designed for smaller-scale experiments where controlling metal ion concentration is critical. EDTA is essential in a variety of fields, including molecular biology, biochemistry, and environmental chemistry, as it prevents metal ions from interfering with biological reactions and analytical procedures. The ability of EDTA to sequester divalent metal ions, particularly calcium and magnesium, makes it indispensable in enzyme assays, protein purification, and DNA/RNA manipulation. It is commonly used in buffer solutions and as a preservative in biological samples.