Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylenediaminetetraacetic acid hydrate sodium | 85715-60-2 | MFCD00149675 | 97.0% | 376.20 g/mol | C10H13N2Na3O8 · xH2O | 100 G
SDP

Catalog No. 5000282740
Encompass_Preferred

Ethylenediaminetetraacetic acid sodium hydrate is a research-grade chelating agent and buffer component used to bind metal ions, stabilize solutions, and support a variety of laboratory protocols. It is supplied as a solid suitable for analytical and preparative applications.

  • High purity (97.0%).
  • Molecular weight: 376.20 g/mol.
  • Chemical formula: C10H13N2Na3O8 · xH2O.
  • Appearance: white to off-white solid.
  • Function: chelating agent for metal ions and buffer formulations.
  • Available in 100 G packaging for laboratory use.

Catalog No. 50-002-82740 Supplier Medchemexpress LLC Supplier No. HYW127774100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylenediaminetetraacetic acid sodium hydrate is a research-grade chelating agent and buffer component used to bind metal ions, stabilize solutions, and support a variety of laboratory protocols. It is supplied as a solid suitable for analytical and preparative applications.

  • High purity (97.0%).
  • Molecular weight: 376.20 g/mol.
  • Chemical formula: C10H13N2Na3O8 · xH2O.
  • Appearance: white to off-white solid.
  • Function: chelating agent for metal ions and buffer formulations.
  • Available in 100 G packaging for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.