Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid magnesium disodium tetrahydrate | 29932-54-5 | MFCD03414198 | 99.0% | 430.56 g/mol | C10H20MgN2Na2O12 | 100 G
SDP
Catalog No. 5000283353
Encompass_Preferred
Magnesium disodium EDTA tetrahydrate is the magnesium salt of ethylenediaminetetraacetic acid used as a chelating reagent in biochemical and analytical applications. It binds divalent and trivalent metal ions, functions as a reversible activator of fructose diphosphatase, and is supplied as a high-purity solid suitable for laboratory assays and metal-ion control.
  • High purity (≈99.0%) suitable for analytical use.
  • Effective chelator of divalent and trivalent metal ions.
  • Reversible activator of fructose diphosphatase in biochemical assays.
  • Solid, easy-to-weigh tetrahydrate form for reproducible results.
  • Available in convenient laboratory pack sizes.
Catalog No. 50-002-83353 Supplier Medchemexpress LLC Supplier No. HYW134005100G
Magnesium disodium EDTA tetrahydrate is the magnesium salt of ethylenediaminetetraacetic acid used as a chelating reagent in biochemical and analytical applications. It binds divalent and trivalent metal ions, functions as a reversible activator of fructose diphosphatase, and is supplied as a high-purity solid suitable for laboratory assays and metal-ion control.
  • High purity (≈99.0%) suitable for analytical use.
  • Effective chelator of divalent and trivalent metal ions.
  • Reversible activator of fructose diphosphatase in biochemical assays.
  • Solid, easy-to-weigh tetrahydrate form for reproducible results.
  • Available in convenient laboratory pack sizes.