Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid sodium hydrate | 85715-60-2 | MFCD00013305 | 97.0% | 376.20 g/mol | C10H13N2Na3O8 · xH2O | 50 G
SDP
Catalog No. 5000282773
Encompass_Preferred
EDTA hydrate sodium is the trisodium salt hydrate form of ethylenediaminetetraacetic acid used as a chelating agent and biochemical reagent in life-science research. Supplied as a white to off-white solid with ~97.0% purity and a molecular weight of 376.20 g/mol, it is commonly used to sequester metal ions, stabilize formulations, and buffer solutions in laboratory applications.
  • Chelates divalent and trivalent metal ions to prevent metal-catalyzed reactions.
  • Stabilizes enzyme preparations and biochemical assays by binding trace metals.
  • Acts as a buffer component in solution preparation and pH control.
  • High-purity solid, easy to weigh and dissolve for reproducible results.
  • Available in common laboratory pack sizes for convenient use.
Catalog No. 50-002-82773 Supplier Medchemexpress LLC Supplier No. HYW12777450G
EDTA hydrate sodium is the trisodium salt hydrate form of ethylenediaminetetraacetic acid used as a chelating agent and biochemical reagent in life-science research. Supplied as a white to off-white solid with ~97.0% purity and a molecular weight of 376.20 g/mol, it is commonly used to sequester metal ions, stabilize formulations, and buffer solutions in laboratory applications.
  • Chelates divalent and trivalent metal ions to prevent metal-catalyzed reactions.
  • Stabilizes enzyme preparations and biochemical assays by binding trace metals.
  • Acts as a buffer component in solution preparation and pH control.
  • High-purity solid, easy to weigh and dissolve for reproducible results.
  • Available in common laboratory pack sizes for convenient use.