Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid sodium hydrate | 85715-60-2 | MFCD00149675 | 97.0% | 376.20 g/mol | C10H15N2Na3O9 | 25 G
SDP
Catalog No. 5000283512
Encompass_Preferred
EDTA sodium hydrate (ethylenediaminetetraacetic acid trisodium salt hydrate) is a sodium salt hydrate of EDTA used as a metal chelating agent and laboratory reagent. It binds divalent and trivalent metal ions to stabilize solutions and remove trace metals during buffer preparation, sample cleanup, and analytical assays. Supplied as a crystalline powder for research use.
  • High purity (97.0%).
  • Effective chelator of Ca2+ and Mg2+.
  • Stable crystalline powder, easy to weigh and dissolve.
  • Suitable for buffer preparation and analytical assays.
  • Available in multiple pack sizes, including 25 g.
  • Identified by CAS number 85715-60-2 for traceability.
Catalog No. 50-002-83512 Supplier Medchemexpress LLC Supplier No. HYW12777425G
EDTA sodium hydrate (ethylenediaminetetraacetic acid trisodium salt hydrate) is a sodium salt hydrate of EDTA used as a metal chelating agent and laboratory reagent. It binds divalent and trivalent metal ions to stabilize solutions and remove trace metals during buffer preparation, sample cleanup, and analytical assays. Supplied as a crystalline powder for research use.
  • High purity (97.0%).
  • Effective chelator of Ca2+ and Mg2+.
  • Stable crystalline powder, easy to weigh and dissolve.
  • Suitable for buffer preparation and analytical assays.
  • Available in multiple pack sizes, including 25 g.
  • Identified by CAS number 85715-60-2 for traceability.