Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylenediaminetetraacetic Acid Tetrasodium Salt Hydrate

Catalog No. 03701250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03-701-250G 250 g
03-701-1KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 03-701-250G Supplier Sigma Aldrich Supplier No. 03701250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

EDTA is an inhibitor of metalloproteases, at effective concentrations of 1-10 mM. EDTA acts as a chelator of the zinc ion in the active site of metalloproteases, and can also inhibit other metal ion-dependent proteases such as calcium-dependent cysteine proteases. For use as an anticoagulant, disodium or tripotassium salts of EDTA are most commonly used. The optimal concentration is 1.5 mg per ml of blood.

Specifications

CAS 194491-31-1
Molecular Formula C10H12N2Na4O8 -+ xH2O
Linear Formula (NaOOCCH2)2NCH2CH2N(CH2COONa)2 -+ xH2O
MDL Number MFCD00150025
Quantity 250 g
Synonym Tetrasodium EDTA; EDTA tetrasodium salt; Edathamil; Tetrasodium ethylenediaminetetraacetate hydrate
Molecular Weight (g/mol) 380.17 g/mol
Biological Activity Respiratory Tract
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.