Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, 99.5%, for molecular biology, DNAse, RNAse and Protease free

Catalog No. ac327211000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC327211000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC327211000 Supplier Thermo Scientific Chemicals Supplier No. 327211000
Only null left
Add to Cart

CAS: 13235-36-4 | C10H20N2Na4O12 | 452.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13235-36-4
Molecular Formula C10H20N2Na4O12
Molecular Weight (g/mol) 452.23
MDL Number MFCD00150027
InChI Key XFLNVMPCPRLYBE-UHFFFAOYSA-J
Synonym tetrasodium ethylenediaminetetraacetate tetrahydrate, edetate sodium tetrahydrate, unii-l13nhd21x6, ethylenediaminetetraacetic acid tetrasodium salt tetrahydrate, acmc-209bp2, ksc910c4l, edta, tetrasodium tetrahydrate salt, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, tetrasodium salt, tetrahydrate, ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate, glycine, n,n'-1,2-ethanediylbis n-carboxymethyl-, sodium salt, hydrate 1:4:4
PubChem CID 23287279
IUPAC Name tetrasodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
SMILES O.O.O.O.[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Absorbance 0.06 max. (0.1 M in water) at 280nm, 1.5 max. (0.1 M in water) at 260nm
Color White
pH 10.7 to 11.7 (0.1M aq. soln.)
Flash Point >100°C
Assay Percent Range 99.4% min. (Complexometry)
Packaging Glass bottle
Quantity 100 g
Fieser 01,373; 05,296
Merck Index 14, 3517
Solubility Information (0.1 M in water) Clear colorless
Formula Weight 452.23
Percent Purity 99.5%
Grade Molecular Biology
Physical Form Crystalline Powder
Chemical Name or Material Ethylenediaminetetraacetic acid, tetrasodium salt tetrahydrate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physi

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.