Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid trisodium salt | 150-38-9 | 98.0% | 500 MG
SDP
Catalog No. 5000407517
Encompass_Preferred
Ethylenediaminetetraacetic acid trisodium salt, also known as EDTA trisodium salt or Trisodium EDTA, is a metal chelating agent that binds to bivalent and trivalent metal cations, including calcium. It exhibits a range of biological activities, making it suitable for various research applications.
  • Binds to bivalent and trivalent metal cations (e.g., calcium)
  • Exhibits antibacterial, anti-inflammatory, antioxidant, anti-hypercalcemia, and anticoagulant activities
  • Decreases metal ion-catalyzed oxidative damage to proteins
  • Maintains a reducing environment during protein purification
  • Alleviates liver fibrosis
  • Used in research for coronary artery disease and neural system diseases
Catalog No. 50-004-07517 Supplier Medchemexpress LLC Supplier No. HYB1009500MG
Ethylenediaminetetraacetic acid trisodium salt, also known as EDTA trisodium salt or Trisodium EDTA, is a metal chelating agent that binds to bivalent and trivalent metal cations, including calcium. It exhibits a range of biological activities, making it suitable for various research applications.
  • Binds to bivalent and trivalent metal cations (e.g., calcium)
  • Exhibits antibacterial, anti-inflammatory, antioxidant, anti-hypercalcemia, and anticoagulant activities
  • Decreases metal ion-catalyzed oxidative damage to proteins
  • Maintains a reducing environment during protein purification
  • Alleviates liver fibrosis
  • Used in research for coronary artery disease and neural system diseases