Learn More
Medchemexpress LLC Ethylenediaminetetraacetic acid zinc disodium tetrahydrate | 176736-49-5 | MFCD00151728 | 95.0% | 471.63 g/mol | C10H20N2Na2O12Zn | 100 G
SDP
Catalog No. 5000282774
Encompass_Preferred
Zn(II)-EDTA disodium tetrahydrate is the disodium tetrahydrate salt of ethylenediaminetetraacetic acid coordinated to zinc, supplied as a reagent-grade powder for use as a zinc chelator and biochemical reagent in life-science research.
  • High-purity reagent (95.0%) suitable for biochemical assays.
  • Zinc chelator useful for metal complexation and enzymatic studies.
  • Supplied as a white to off-white powder tetrahydrate for easy weighing.
  • Molecular weight 471.63 g/mol and CAS 176736-49-5 for identification.
  • Available in laboratory-scale pack sizes, including 100 G.
  • Stable when stored as powder at -20°C (3 years) or 4°C (2 years).
Catalog No. 50-002-82774 Supplier Medchemexpress LLC Supplier No. HYW127776100G
Zn(II)-EDTA disodium tetrahydrate is the disodium tetrahydrate salt of ethylenediaminetetraacetic acid coordinated to zinc, supplied as a reagent-grade powder for use as a zinc chelator and biochemical reagent in life-science research.
  • High-purity reagent (95.0%) suitable for biochemical assays.
  • Zinc chelator useful for metal complexation and enzymatic studies.
  • Supplied as a white to off-white powder tetrahydrate for easy weighing.
  • Molecular weight 471.63 g/mol and CAS 176736-49-5 for identification.
  • Available in laboratory-scale pack sizes, including 100 G.
  • Stable when stored as powder at -20°C (3 years) or 4°C (2 years).