Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ethylenedinitrilo)tetraacetic Acid, Powder, ACS, 99.4-100.6%, Spectrum™ Chemical
SDP

Catalog No. 18608015 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-015 45 kg Poly Bottle
18-608-011 125 g Poly Bottle
18-608-012 500 g Poly Bottle
18-608-013 2.5 kg Poly Bottle
18-608-014 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-608-015 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E104045KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
IUPAC Name 2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

CAS Max % 1
Ignition Residue 0.002
Packaging Poly Bottle
Quantity 45 kg
Percent Purity 99.4 to 100.6%
Grade ACS
Physical Form Crystalline Powder or Lumps
Chemical Name or Material (Ethylenedinitrilo)tetraacetic Acid, Powder

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.