Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ethylenedinitrilo)tetraacetic Acid, Powder, Spectrum™ Chemical
SDP

Catalog No. 18620739 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-739 45 kg
18-620-735 125 g
18-620-736 500 g
18-620-737 2.5 kg
18-620-738 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-620-739 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E353245KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synonym: EDTA, Edetic Acid, Ethylenediaminetetraacetic Acid, Edathamil

Chemical Identifiers

CAS 60-00-4

Specifications

pH 2.0 to 3.0 (1% W/V dilution)
Physical Form Powder
Quantity 45 kg
Packaging Drum
Molecular Weight (g/mol) 292.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.