Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethylenediaminetetraacetic acid disodium salt dihydrate, 6381-92-6, MFCD00150037, 500 g
Catalog No. NC1308047
Encompass
Empirical Formula (Hill Notation): C10H14N2Na2O8 · 2H2O, Molecular Weight: 372.24, ACS reagent, 99.0-101.0%, Synonym: Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2. Ethylenediaminetetraacetic acid (EDTA) is an anticoagulant, majorly used in preventing clotting of blood.
Catalog No. NC1308047 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. E4884500G
Empirical Formula (Hill Notation): C10H14N2Na2O8 · 2H2O, Molecular Weight: 372.24, ACS reagent, 99.0-101.0%, Synonym: Disodium ethylenediaminetetraacetate dihydrate, EDTA disodium salt, EDTA-Na2, Edathamil, Edetate disodium salt dihydrate, Sequestrene Na2. Ethylenediaminetetraacetic acid (EDTA) is an anticoagulant, majorly used in preventing clotting of blood.