Learn More
Medchemexpress LLC Ethylenethiourea-d4 | 352431-28-8 | 106.18 | 1 MG
SDP
Catalog No. 5000452308
Encompass_Preferred
Ethylenethiourea-d4 is the deuterium labeled Ethylenethiourea. Ethylenethiourea is a degradation product of the ethylenebisthiocarbamate group of fungicides. Ethylenethiourea is tumorigenic and teratogenic, and it is orally active.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-52308 Supplier Medchemexpress LLC Supplier No. HYW010593S1MG
Ethylenethiourea-d4 is the deuterium labeled Ethylenethiourea. Ethylenethiourea is a degradation product of the ethylenebisthiocarbamate group of fungicides. Ethylenethiourea is tumorigenic and teratogenic, and it is orally active.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.