Learn More
Medchemexpress LLC Ethylenethiourea-d4 | 352431-28-8 | 106.18 | 10 MG
SDP
Catalog No. 5000366532
Encompass_Preferred
Ethylenethiourea-d4 is a deuterium-labeled Ethylenethiourea, which is a degradation product of ethylenebisthiocarbamate fungicides. It has been noted for its tumorigenic and teratogenic properties and is orally active.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-66532 Supplier Medchemexpress LLC Supplier No. HYW010593S10MG
Ethylenethiourea-d4 is a deuterium-labeled Ethylenethiourea, which is a degradation product of ethylenebisthiocarbamate fungicides. It has been noted for its tumorigenic and teratogenic properties and is orally active.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution can affect the pharmacokinetic and metabolic profiles of drugs.