Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylhexadecyldimethylammonium Bromide, High Purity, 99%, Spectrum™ Chemical
SDP

Catalog No. 18610459 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
12 kg
25 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-459 25 kg Amber Glass Bottle
18-610-454 5 g Amber Glass Bottle
18-610-455 100 g Amber Glass Bottle
18-610-456 500 g Amber Glass Bottle
18-610-457 2.5 kg Amber Glass Bottle
18-610-458 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-610-459 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E105425KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 124-03-8

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 kg
Percent Purity 0.99
Grade High Purity
Physical Form Powder or Waxy Solid
Chemical Name or Material Ethylhexadecyldimethylammonium Bromide

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.