Learn More
Medchemexpress LLC Ethylhexyl triazone | 88122-99-0 | 98.0% | 823.07 | 10 MG
SDP
Catalog No. 5000344635
Encompass_Preferred
Ethylhexyl triazone is an approved ultraviolet-B (UV-B) chemical filter commonly used in commercial sunscreens. Its large molecular weight suggests it is unlikely to penetrate the skin, which helps in reducing potential associated effects.
  • Approved UV-B chemical filter
  • Unlikely to penetrate the skin
  • Broad absorption profile in the UV-B region
  • Maximum absorption at approximately 311 nm in dioxane and 313 nm in methanol
Catalog No. 50-003-44635 Supplier Medchemexpress LLC Supplier No. HY10965510MM/1ML
Ethylhexyl triazone is an approved ultraviolet-B (UV-B) chemical filter commonly used in commercial sunscreens. Its large molecular weight suggests it is unlikely to penetrate the skin, which helps in reducing potential associated effects.
  • Approved UV-B chemical filter
  • Unlikely to penetrate the skin
  • Broad absorption profile in the UV-B region
  • Maximum absorption at approximately 311 nm in dioxane and 313 nm in methanol