Learn More
Medchemexpress LLC Ethylhexyl triazone | 88122-99-0 | 98.0% | 823.07 | 50 MG
SDP
Catalog No. 5000345046
Encompass_Preferred
Ethylhexyl triazone, also known as Octyl triazone, is a highly effective ultraviolet-B (UV-B) chemical filter widely utilized in commercial sunscreen formulations. Its large molecular weight of 823.07 g/mol suggests minimal skin penetration, thus reducing potential dermal effects. This compound is notable for its broad absorption spectrum within the UV-B region, with peak absorption observed around 311 nm in dioxane and 313 nm in methanol.
  • Approved as a UV-B chemical filter for commercial sunscreens.
  • Large molecular weight, indicating low skin penetration.
  • Exhibits a broad absorption profile in the UV-B range.
  • Maximum absorption at approximately 311 nm (dioxane) and 313 nm (methanol).
Catalog No. 50-003-45046 Supplier Medchemexpress LLC Supplier No. HY109655500MG
Ethylhexyl triazone, also known as Octyl triazone, is a highly effective ultraviolet-B (UV-B) chemical filter widely utilized in commercial sunscreen formulations. Its large molecular weight of 823.07 g/mol suggests minimal skin penetration, thus reducing potential dermal effects. This compound is notable for its broad absorption spectrum within the UV-B region, with peak absorption observed around 311 nm in dioxane and 313 nm in methanol.
  • Approved as a UV-B chemical filter for commercial sunscreens.
  • Large molecular weight, indicating low skin penetration.
  • Exhibits a broad absorption profile in the UV-B range.
  • Maximum absorption at approximately 311 nm (dioxane) and 313 nm (methanol).