Learn More
Medchemexpress LLC Ethylhexylglycerin 1kg
SDP
Catalog No. 5000204857
Encompass_Preferred
Ethylhexylglycerin has antimicrobial activity inhibiting the growth of Acinetobacter spp Pseudomonas aeruginosa complex and Stenotrophomonas maltophilia Ethylhexylglycerin can cause pigmentary contact dermatitis[1][2]
Catalog No. 50-002-04857 Supplier Medchemexpress LLC Supplier No. HYW0997291KG
Ethylhexylglycerin has antimicrobial activity inhibiting the growth of Acinetobacter spp Pseudomonas aeruginosa complex and Stenotrophomonas maltophilia Ethylhexylglycerin can cause pigmentary contact dermatitis[1][2]