Learn More
TARGETMOL CHEMICALS INC ETHYLHYDROCUPREINE HYDRO 50MG
SDP
Catalog No. 502579992
Encompass_Preferred
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ethylhydrocupreine hydrochloride (Optochin hydrochloride) (Optochin hydrochloride) is a quinine derivate with antimicrobial activity against S. pneumoniae. Ethylhydrocupreine hydrochloride also possesses antimalarial activity against Plasmodium falciparum with an IC50 of 25.75 nM. Ethylhydrocupreine hydrochloride is an agonist of Gallus gallus taste 2 receptors ( ggTas2r1 ggTas2r2 and ggTas2r7 ). purity: 99%
Catalog No. 50-257-9992 Supplier TARGETMOL CHEMICALS INC Supplier No. T4041750MG
Also available in 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ethylhydrocupreine hydrochloride (Optochin hydrochloride) (Optochin hydrochloride) is a quinine derivate with antimicrobial activity against S. pneumoniae. Ethylhydrocupreine hydrochloride also possesses antimalarial activity against Plasmodium falciparum with an IC50 of 25.75 nM. Ethylhydrocupreine hydrochloride is an agonist of Gallus gallus taste 2 receptors ( ggTas2r1 ggTas2r2 and ggTas2r7 ). purity: 99%