Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethyllithium Solution

Catalog No. 561452100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
56-145-2100ML 100 mL
56-145-225ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 56-145-2100ML Supplier Sigma Aldrich Supplier No. 561452100ML
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethyllithium is a general organolithium reagent used in the nucleophilic addition reaction to synthesize a variety of intermediates.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 811-49-4
Density 0.862 g/mL at 25°C
Molecular Formula C2H5Li
Linear Formula CH3CH2Li
MDL Number MFCD00053747
Quantity 100 mL
Molecular Weight (g/mol) 36 g/mol
Biological Activity Blood
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.