Learn More
Strem, An Ascensus Company CAS# 925-90-6. 0.25mole. Ethylmagnesium bromide, 3.4M (40wt% ±1wt%) in 2-methyltetrahydrofuran. MFCD00000043
SDP
Supplier:  Strem, An Ascensus Company 120810250MMOL
Encompass
CAS# 925-90-6. 0.25mole. Ethylmagnesium bromide, 3.4M (40wt% ±1wt%) in 2-methyltetrahydrofuran. MFCD00000043. Molecular Weight: 133.28. Molecular Formula: C2H5BrMg. Color/form: dark brown liq. Strem# 12-0810. http://www.strem.com/catalog/v/12-0810/
Catalog No. 50-367-2