Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylmagnesium Bromide Solution

Catalog No. 36467325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-467-325ML 4 x 25 mL
36-467-3800ML 800 mL
36-467-3100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 36-467-325ML Supplier Sigma Aldrich Supplier No. 3646734X25ML
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylmagnesium bromide solution can be used in Grignard reaction and to synthesize other Grignard reagents.Storage below 25°C may cause formation of crystalline magnesium salts. Moving container to a war mLocation and occasional gentle swirling should redissolve the solid.

Specifications

CAS 925-90-6
Density 1.010 g/mL at 25°C
Molecular Formula C2H5BrMg
Linear Formula CH3CH2MgBr
MDL Number MFCD00000043
Quantity 4 x 25 mL
Synonym EtMgBr solution
Molecular Weight (g/mol) 133.27 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.