Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethylmagnesium Bromide Solution

Catalog No. 1898718L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
4 x 25 mL
800 mL
8 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-987-18L 8 L
18-987-1100ML 100 mL
18-987-1800ML 800 mL
18-987-125ML 4 x 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Danger
Catalog No. 18-987-18L Supplier Sigma Aldrich Supplier No. 1898718L
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethylmagnesium bromide, an organomagnesium compound, is commonly known as Grignard reagent. It is a powerful carbon nucleophile mainly used for C-C bond formation.Ethylmagnesium bromide solution (3.0 M in diethyl ether) can be used for copper(I)-catalyzed allylic substitution reaction.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 925-90-6
Density 1.02 g/mL at 25°C
Molecular Formula C2H5BrMg
Linear Formula CH3CH2MgBr
MDL Number MFCD00000043
Quantity 8 L
Synonym EtMgBr solution
Molecular Weight (g/mol) 133.27 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.