Learn More
Cayman Chemical Ethylmalonyl Coenzyme A (sodium salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502995243
Encompass_Preferred
An intermediate in the ethylmalonyl-CoA pathway in bacteria
Catalog No. 50-299-5243 Supplier Cayman Chemical Supplier No. 235031 MG
An intermediate in the ethylmalonyl-CoA pathway in bacteria