Learn More
Cayman Chemical EthylmaLNyl Coenzyme AsodI
SDP
Catalog No. 502995346
Encompass_Preferred
An intermediate in the ethylmalonyl-CoA pathway in bacteria
Catalog No. 50-299-5346 Supplier Cayman Chemical Supplier No. 23503500 UG
An intermediate in the ethylmalonyl-CoA pathway in bacteria