Learn More
Apexbio Technology LLC ETHYLMALONIC-ACID-1G
SDP
Catalog No. 5000568844
Encompass_Preferred
Ethylmalonic acid (CAS No 601-75-2) is a dicarboxylic acid frequently studied in the field of biomedical research due to its role in metabolic pathways Originating from the breakdown of certain fatty acids it is involved in the examination of metabolic disorders particularly ethylmalonic encephalopathy Ethylmalonic acid acts by interfering with normal fatty acid oxidation processes and mitochondrial function Research often utilizes cell culture models and animal studies to explore its effects with experimental concentrations typically depending on the specific research design Common in vitro activity is observed in the nanomolar to low micromolar range It is used to screen compounds that might act on metabolic targets aiding in the development of therapeutic strategies for related conditions
Catalog No. 50-005-68844 Supplier Apexbio Technology LLC Supplier No. C66021000
Add to Cart
Add to Cart
Ethylmalonic acid (CAS No 601-75-2) is a dicarboxylic acid frequently studied in the field of biomedical research due to its role in metabolic pathways Originating from the breakdown of certain fatty acids it is involved in the examination of metabolic disorders particularly ethylmalonic encephalopathy Ethylmalonic acid acts by interfering with normal fatty acid oxidation processes and mitochondrial function Research often utilizes cell culture models and animal studies to explore its effects with experimental concentrations typically depending on the specific research design Common in vitro activity is observed in the nanomolar to low micromolar range It is used to screen compounds that might act on metabolic targets aiding in the development of therapeutic strategies for related conditions