Learn More
Medchemexpress LLC Ethylmalonic acid-d3 | 70907-93-6 | 98.0% | 135.13 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY34740S5MG
Encompass_Preferred
Ethylmalonic acid-d3 is the deuterium labeled Ethylmalonic acid. It is a short-chain organic dicarboxylic acid that synergistically induces mitochondrial permeability transition (MP) with Ca2+, inhibits Mi-CK, and disrupts mitochondrial energy metabolism. This compound is suitable for research into SCADD, EE, and other genetic metabolic diseases characterized by EMA accumulation.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Inhibits Mi-CK
  • Disrupts mitochondrial energy metabolism
  • Deuterium labeled version of ethylmalonic acid
Catalog No. 50-004-03838