Learn More
Medchemexpress LLC Ethylmalonic acid-d5 | 66311-22-6 | 98.1% | 137.15 | 1 MG
SDP
Catalog No. 5000404312
Encompass_Preferred
Ethylmalonic acid-d5 is a deuterium labeled short-chain organic dicarboxylic acid. It synergistically induces mitochondrial permeability transition with Ca2+, inhibits Mi-CK, and disrupts mitochondrial energy metabolism. This compound can be used in the research of SCADD, EE, and other genetic metabolic diseases characterized by EMA accumulation.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound for pharmacokinetic and metabolic studies
  • Inhibits mitochondrial creatine kinase activity
  • Disrupts mitochondrial energy metabolism
Catalog No. 50-004-04312 Supplier Medchemexpress LLC Supplier No. HY34740S11MG
Ethylmalonic acid-d5 is a deuterium labeled short-chain organic dicarboxylic acid. It synergistically induces mitochondrial permeability transition with Ca2+, inhibits Mi-CK, and disrupts mitochondrial energy metabolism. This compound can be used in the research of SCADD, EE, and other genetic metabolic diseases characterized by EMA accumulation.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled compound for pharmacokinetic and metabolic studies
  • Inhibits mitochondrial creatine kinase activity
  • Disrupts mitochondrial energy metabolism