Learn More
Medchemexpress LLC Ethylparaben | 120-47-8 | 10 G
SDP
Supplier:  Medchemexpress LLC HYB093410G
Encompass_Preferred
Ethylparaben is the ethyl ester of paraben, commonly utilized as an antifungal preservative and food additive. This substance is provided as a solid and is primarily intended for laboratory chemical applications and the manufacture of various substances, requiring handling by experienced personnel in appropriately equipped facilities.
  • Used as an antifungal preservative and food additive
  • Appears as a solid
  • Stable under recommended storage conditions
  • For research use only
  • Toxic to aquatic life
Catalog No. 50-004-07881