Learn More
U.S. Pharmacopeia Ethylparaben, 120-47-8, MFCD00002353, 200mg
Catalog No. NC9983583
Encompass
Molecular formula C9H10O3, Molecular weight 166.17, Melting Point 239 - 244.4 °F (115 - 118 °C), Boiling Point 566.6 - 568.4 °F (297 - 298 °C) (decomposes), Synonyms: Ethyl p-hydroxybenzoate
Catalog No. NC9983583 Supplier U.S. Pharmacopeia Supplier No. 1267000
Molecular formula C9H10O3, Molecular weight 166.17, Melting Point 239 - 244.4 °F (115 - 118 °C), Boiling Point 566.6 - 568.4 °F (297 - 298 °C) (decomposes), Synonyms: Ethyl p-hydroxybenzoate