Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethylparaben-d4 | 1219795-53-5 | 99.6% | 5 MG
SDP

Catalog No. 5000409106
Encompass_Preferred

Ethylparaben-d4 is a deuterium-labeled Ethylparaben intended for research use only. This compound can function as a tracer or an internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS. It exhibits high purity and specific storage requirements to maintain its stability.

  • High purity of 99.6%
  • Molecular weight of 170.20
  • Solid, white to off-white appearance
  • Store powder at -20°C for up to 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)

Catalog No. 50-004-09106 Supplier Medchemexpress LLC Supplier No. HYB0934S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethylparaben-d4 is a deuterium-labeled Ethylparaben intended for research use only. This compound can function as a tracer or an internal standard for quantitative analysis using methods like NMR, GC-MS, or LC-MS. It exhibits high purity and specific storage requirements to maintain its stability.

  • High purity of 99.6%
  • Molecular weight of 170.20
  • Solid, white to off-white appearance
  • Store powder at -20°C for up to 3 years
  • Store in solvent at -80°C for 6 months, or -20°C for 1 month
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis (NMR, GC-MS, LC-MS)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.