Learn More
Cayman Chemical Ethyl (R)-(-)-3-Hydroxybutyrate, Purity: ≥95%, CAS: 24915-95-5, 25 g
SDP
Catalog No. 502983302
Encompass_Preferred
A chiral starting material for the production of numerous biologically active compounds
Catalog No. 50-298-3302 Supplier Cayman Chemical Supplier No. 2065925 G
A chiral starting material for the production of numerous biologically active compounds