Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ethylthio)acetone, 95%

Catalog No. AAL0414206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0414206 5 g
AAL0414214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0414206 Supplier Thermo Scientific Chemicals Supplier No. L0414206
Add to Cart

CAS: 20996-62-7 | C5H10OS | 118.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20996-62-7
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00026976
InChI Key ZKDXKBIOEVBFGV-UHFFFAOYSA-N
PubChem CID 88750
IUPAC Name 1-ethylsulfanylpropan-2-one
SMILES CCSCC(=O)C

Specifications

Boiling Point 170°C to 172°C
Flash Point 52°C (125°F)
Refractive Index 1.472
Quantity 5 g
UN Number UN1993
Beilstein 1740830
Synonym ethylthio acetone, 2-propanone, 1-ethylthio, 1-ethylthio acetone, ethyl 2-oxopropyl sulfide, 1-ethylthio-2-propanone, 2-propanone,1-ethylthio, 1-ethylsulfanyl propan-2-one, thioethoxy acetone, acmc-1cazk, ch3coch2sch2ch3
Formula Weight 118.2
Percent Purity 95%
Chemical Name or Material (Ethylthio)acetone
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 244-143-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.