Learn More
Strem, An Ascensus Company CAS# 383-63-1. 25g. Ethyltrifluoroacetate, 99%. MFCD00000419
SDP
Catalog No. 5090109303
Encompass
CAS# 383-63-1. 25g. Ethyltrifluoroacetate, 99%. MFCD00000419. Molecular Weight: 142.08. Molecular Formula: CF3CO2C2H5. Color/form: colorless liq. Strem# 09-3270. http://www.strem.com/catalog/v/09-3270/
Catalog No. 50-901-09303 Supplier Strem, An Ascensus Company Supplier No. 09327025G
CAS# 383-63-1. 25g. Ethyltrifluoroacetate, 99%. MFCD00000419. Molecular Weight: 142.08. Molecular Formula: CF3CO2C2H5. Color/form: colorless liq. Strem# 09-3270. http://www.strem.com/catalog/v/09-3270/